siRNA Details
| Gene Symbol: OSBPL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242508.1 | NP_001229437.1 | 13 | 1423 | ||
| NM_018030.4 | NP_060500.3 | 11 | 1792 | |||
| NM_080597.3 | NP_542164.2 | 25 | 2679 | |||
| XM_006722380.3 | XP_006722443.1 | 24 | 2592 | |||
| XM_006722382.3 | XP_006722445.1 | 24 | 2557 | |||
| XM_017025530.1 | XP_016881019.1 | 25 | 2971 | |||
| XM_017025531.1 | XP_016881020.1 | 23 | 2281 | |||
| XM_017025533.1 | XP_016881022.1 | 11 | 1275 | |||
| XM_017025534.1 | XP_016881023.1 | 11 | 3803 | |||
| GenBank mRNA | AF274714.1 | - | 11 | 1792 | ||
| AF323726.2 | - | 25 | 2567 | |||
| AF392449.1 | - | 11 | 1413 | |||
| AF392450.1 | - | 25 | 2636 | |||
| AK096358.1 | - | 25 | 2679 | |||
| AK292895.1 | - | 26 | 2611 | |||
| AK301862.1 | - | 13 | 1423 | |||
| AK314552.1 | - | 11 | 1243 | |||
| BC063420.1 | - | 11 | 1478 | |||
| BC073854.1 | - | 9 | 999 | |||
| BC098384.1 | - | 11 | 1471 | |||
| BC136452.1 | - | 25 | 2576 | 9020441 | ||
| BC144117.1 | - | 25 | 2576 | 9052637 | ||

OSBPL1A
Genomic Map