siRNA Details
| Gene Symbol: C1QTNF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030945.3 | NP_112207.1 | 2 | 266 | ||
| NM_181435.5 | NP_852100.3 | 2 | 485 | |||
| GenBank mRNA | AF173888.1 | - | 2 | 168 | ||
| AF326976.1 | - | 2 | 201 | |||
| AF329837.1 | - | 2 | 208 | |||
| AK075533.1 | - | 2 | 213 | |||
| AK295968.1 | - | 2 | 485 | |||
| AK315921.1 | - | 2 | 472 | |||
| AY358388.1 | - | 2 | 201 | |||
| BC112925.1 | - | 2 | 140 | 5199982 | ||
| BC120990.2 | - | 2 | 415 | 40116366 | ||
| BX483406.1 | - | 2 | 432 | |||
| BX640995.1 | - | 2, 3 | 232 | |||
| EU399231.1 | - | 2 | 130 | |||
| EU399232.1 | - | 2 | 349 | |||
| Gene Symbol: C1QTNF3-AMACR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037951.1 | 2 | 152 | |||
| GenBank mRNA | AB209261.1 | - | 2 | 161 | ||

C1QTNF3
Genomic Map