siRNA Details
| Gene Symbol: C1QTNF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031910.3 | NP_114116.3 | 3 | 585 | ||
| NM_182486.1 | NP_872292.1 | 3 | 585 | |||
| XM_011529857.1 | XP_011528159.1 | 4 | 3442 | |||
| XM_017028569.1 | XP_016884058.1 | 3 | 2734 | |||
| XM_017028570.1 | XP_016884059.1 | 5 | 1352 | |||
| XM_017028571.1 | XP_016884060.1 | 5 | 3090 | |||
| XM_017028572.1 | XP_016884061.1 | 5 | 3079 | |||
| XM_017028573.1 | XP_016884062.1 | 5 | 868 | |||
| XM_017028574.1 | XP_016884063.1 | 4 | 661 | |||
| GenBank mRNA | AF329842.1 | - | 3 | 523 | ||
| AK128125.1 | - | 8 | 3937 | |||
| AK226149.1 | - | 2 | 1108 | |||
| AK314060.1 | - | 3 | 542 | |||
| AY358374.1 | - | 5 | 722 | |||
| BC020551.1 | - | 3 | 584 | 2900683 | ||
| BU147265.1 | - | 3 | 587 | |||
| BU535692.1 | - | 2 | 171 | |||
| CB961508.1 | - | 3 | 569 | |||
| CR456451.1 | - | 3 | 535 | |||

C1QTNF6
Genomic Map