siRNA Details
| Gene Symbol: C1QTNF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135170.1 | NP_001128642.1 | 3 | 924 | ||
| NM_001135171.1 | NP_001128643.1 | 3 | 871 | |||
| NM_031911.4 | NP_114117.1 | 3 | 754 | |||
| XM_011513772.1 | XP_011512074.1 | 4 | 794 | |||
| XM_011513773.1 | XP_011512075.1 | 4 | 843 | |||
| GenBank mRNA | AF329839.1 | - | 3 | 729 | ||
| AK314802.1 | - | 3 | 872 | |||
| BC022187.2 | - | 3 | 872 | 4825040 | ||
| BC024015.2 | - | 3 | 872 | 4823626 | ||
| BX647781.1 | - | 3 | 924 | |||

C1QTNF7
Genomic Map