siRNA Details
| Gene Symbol: ZNF618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318040.1 | NP_001304969.1 | 3, 4 | 444 | ||
| NM_001318041.1 | NP_001304970.1 | 3, 4 | 444 | |||
| NM_133374.2 | NP_588615.2 | 3, 4 | 444 | |||
| XM_011518203.2 | XP_011516505.1 | 3, 4 | 429 | |||
| XM_011518204.2 | XP_011516506.1 | 3, 4 | 429 | |||
| XM_017014245.1 | XP_016869734.1 | 3, 4 | 429 | |||
| XM_017014246.1 | XP_016869735.1 | 3, 4 | 429 | |||
| GenBank mRNA | AK126896.1 | - | 3, 4 | 385 | ||
| BC053892.1 | - | 3, 4 | 377 | |||
| BC136287.1 | - | 3, 4 | 392 | 9020274 | ||

ZNF618
Genomic Map