siRNA Details
| Gene Symbol: HAUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138443.3 | NP_612452.1 | 7 | 803 | ||
| NR_026978.1 | 7 | 843 | ||||
| GenBank mRNA | AK024747.1 | - | 4 | 266 | ||
| AK097403.1 | - | 7 | 784 | |||
| AK315605.1 | - | 7 | 749 | |||
| AY360137.1 | - | 7 | 803 | |||
| BC005958.1 | - | 7 | 779 | 4047707 | ||
| BC014003.2 | - | 7 | 776 | 3844806 | ||

HAUS1
Genomic Map