siRNA Details
| Gene Symbol: RAB42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193532.1 | NP_001180461.1 | 2 | 1024 | ||
| NM_152304.1 | NP_689517.1 | 2 | 974 | |||
| XM_017000226.1 | XP_016855715.1 | 2 | 1045 | |||
| GenBank mRNA | BC033175.1 | - | 2 | 974 | 4561064 | |

RAB42
Genomic Map