siRNA Details
| Gene Symbol: ZNF501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258280.1 | NP_001245209.1 | 3 | 810 | ||
| NM_145044.3 | NP_659481.2 | 3 | 517 | |||
| GenBank mRNA | AK054613.1 | - | 3 | 514 | ||
| AK297216.1 | - | 3 | 499 | |||
| AK316574.1 | - | 3 | 517 | |||
| BC013762.1 | - | 3 | 492 | 4521303 | ||
| BP368765.1 | - | 3 | 517 | |||

ZNF501
Genomic Map