siRNA Details
| Gene Symbol: FAM46B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052943.3 | NP_443175.2 | 2 | 746 | ||
| GenBank mRNA | AK122816.1 | - | 2 | 876 | ||
| BC012790.2 | - | 2 | 740 | 3957388 | ||
| BC014160.2 | - | 2 | 740 | 4542896 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM46B| Gene Symbol: FAM46B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052943.3 | NP_443175.2 | 2 | 746 | ||
| GenBank mRNA | AK122816.1 | - | 2 | 876 | ||
| BC012790.2 | - | 2 | 740 | 3957388 | ||
| BC014160.2 | - | 2 | 740 | 4542896 | ||