siRNA Details
| Gene Symbol: NOSTRIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039724.3 | NP_001034813.2 | 8 | 641 | ||
| NM_001171631.1 | NP_001165102.1 | 12 | 1342 | |||
| NM_001171632.1 | NP_001165103.1 | 7 | 557 | |||
| NM_052946.3 | NP_443178.2 | 7 | 548 | |||
| XM_005246270.1 | XP_005246327.1 | 7 | 529 | |||
| XM_006712233.3 | XP_006712296.1 | 8 | 592 | |||
| XM_011510540.2 | XP_011508842.1 | 8 | 592 | |||
| XM_017003278.1 | XP_016858767.1 | 8 | 592 | |||
| XM_017003279.1 | XP_016858768.1 | 7 | 915 | |||
| XM_017003280.1 | XP_016858769.1 | 6 | 407 | |||
| GenBank mRNA | AJ532842.1 | - | 8 | 584 | ||
| AK002203.2 | - | 8 | 613 | |||
| AK093444.1 | - | 12 | 1340 | |||
| AK290254.1 | - | 8 | 607 | |||
| BC014189.2 | - | 7 | 548 | 4650302 | ||
| BC093072.1 | - | 8 | 588 | 30327154 | ||
| DQ402497.1 | - | 7 | 500 | |||

NOSTRIN
Genomic Map