siRNA Details
| Gene Symbol: TSR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001410.2 | NP_001001410.1 | 4 | 651 | ||
| GenBank mRNA | BC013943.1 | - | 4 | 655 | 4064325 | |
| BC016699.1 | - | 4 | 655 | 4097703 | ||
| BC017325.2 | - | 4 | 635 | 4549940 | ||

TSR3
Genomic Map