siRNA Details
| Gene Symbol: CCDC151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302453.1 | NP_001289382.1 | 2 | 186 | ||
| NM_001302454.1 | NP_001289383.1 | 2 | 480 | |||
| NM_145045.4 | NP_659482.3 | 2 | 480 | |||
| XM_017026241.1 | XP_016881730.1 | 2 | 478 | |||
| GenBank mRNA | AK056363.1 | - | 2 | 401 | ||
| AK294355.1 | - | 4 | 380 | |||
| AK302113.1 | - | 2 | 187 | |||
| AK308072.1 | - | 2 | 442 | |||
| BC014252.2 | - | 1 | 37 | |||
| BC141828.1 | - | 2 | 412 | 40148425 | ||
| BC142637.1 | - | 2 | 425 | 40147788 | ||
| BI548500.1 | - | 2 | 445 | |||
| DB040667.1 | - | 2 | 480 | |||

CCDC151
Genomic Map