siRNA Details
| Gene Symbol: RNF166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171815.1 | NP_001165286.1 | 4, 5 | 473 | ||
| NM_001171816.1 | NP_001165287.1 | 5, 6 | 1322 | |||
| NM_178841.3 | NP_849163.1 | 5, 6 | 745 | |||
| XM_017022913.1 | XP_016878402.1 | 5, 6 | 1976 | |||
| GenBank mRNA | AK057106.1 | - | 5, 6 | 1322 | ||
| AK057201.1 | - | 4, 5 | 393 | |||
| AK290920.1 | - | 5, 6 | 712 | |||
| BC013948.2 | - | 5, 6 | 671 | 3509621 | ||
| BC017226.1 | - | 5, 6 | 723 | 4299817 | ||
| BI911983.1 | - | 4, 5 | 480 | |||

RNF166
Genomic Map