siRNA Details
| Gene Symbol: LYSMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286812.1 | NP_001273741.1 | 3 | 590 | ||
| NM_198273.2 | NP_938014.1 | 3 | 649 | |||
| XM_005248421.4 | XP_005248478.1 | 3 | 1114 | |||
| GenBank mRNA | AK291478.1 | - | 3 | 646 | ||
| AL137315.1 | - | 1 | 146 | |||
| AL832721.1 | - | 3 | 630 | |||
| BC058027.1 | - | 3 | 590 | |||
| BC136740.1 | - | 3 | 541 | 9020730 | ||
| BC136741.1 | - | 3 | 541 | 9020731 | ||
| BX537972.1 | - | 3 | 619 | |||

LYSMD3
Genomic Map