siRNA Details
| Gene Symbol: WDR92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256476.1 | NP_001243405.1 | 5 | 726 | ||
| NM_138458.3 | NP_612467.1 | 5 | 726 | |||
| NR_046234.1 | 5 | 599 | ||||
| GenBank mRNA | AK056303.1 | - | 5 | 690 | ||
| AK058090.1 | - | 5 | 599 | |||
| AL834142.1 | - | 5 | 562 | |||
| AY541587.1 | - | 5 | 701 | |||
| BC014022.2 | - | 5 | 648 | 4309224 | ||
| BC020271.1 | - | 5 | 680 | |||
| BC036904.1 | - | 5 | 536 | |||
| BC066657.1 | - | 5 | 676 | 6164689 | ||

WDR92
Genomic Map