siRNA Details
| Gene Symbol: WDR92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138458.3 | NP_612467.1 | 8 | 999 | ||
| GenBank mRNA | AK056303.1 | - | 8 | 963 | ||
| AK058090.1 | - | 8 | 872 | |||
| AK090527.1 | - | 1 | 402 | |||
| AL834142.1 | - | 8 | 3623 | |||
| AY541587.1 | - | 8 | 975 | |||
| BC020271.1 | - | 8 | 953 | |||
| BC066657.1 | - | 8 | 949 | 6164689 | ||

WDR92
Genomic Map