siRNA Details
| Gene Symbol: NUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138459.3 | NP_612468.1 | 3 | 878 | ||
| GenBank mRNA | AK125119.1 | - | 3 | 840 | ||
| AK126335.1 | - | 4 | 592 | |||
| AK130093.1 | - | 2 | 195 | |||
| BC013026.2 | - | 3 | 825 | 3531630 | ||
| BC063794.1 | - | 3 | 832 | 6500832 | ||
| BC066910.1 | - | 3 | 872 | 4830860 | ||
| BC110325.1 | - | 3 | 834 | 5553827 | ||
| BC150654.1 | - | 3 | 845 | 9057025 | ||
| BC150655.1 | - | 3 | 845 | 9057026 | ||

NUS1
Genomic Map