siRNA Details
| Gene Symbol: CYYR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052954.3 | NP_443186.2 | 3, 4 | 796 | ||
| GenBank mRNA | AF401639.1 | - | 3, 4 | 401 | ||
| AK054581.1 | - | 3, 4 | 676 | |||
| AL833200.1 | - | 3, 4 | 607 | |||
| AY061853.1 | - | 3, 4 | 341 | |||
| BC036761.1 | - | 3, 4 | 584 | 5588436 | ||
| JF693914.1 | - | 2, 3 | 269 | |||
| KF154022.1 | - | 4, 5 | 568 | |||

CYYR1
Genomic Map