siRNA Details
| Gene Symbol: TGM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052955.2 | NP_443187.1 | 6 | 930 | ||
| XM_017021903.1 | XP_016877392.1 | 6 | 928 | |||
| GenBank mRNA | AF363393.1 | - | 6 | 930 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TGM7| Gene Symbol: TGM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052955.2 | NP_443187.1 | 6 | 930 | ||
| XM_017021903.1 | XP_016877392.1 | 6 | 928 | |||
| GenBank mRNA | AF363393.1 | - | 6 | 930 | ||