siRNA Details
| Gene Symbol: ZMYND19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138462.2 | NP_612471.1 | 3 | 346 | ||
| XM_005266052.4 | XP_005266109.1 | 3 | 775 | |||
| XM_017014255.1 | XP_016869744.1 | 3 | 779 | |||
| GenBank mRNA | AJ298882.2 | - | 3 | 354 | ||
| AX766496.1 | - | 3 | 354 | |||
| BC012948.1 | - | 3 | 309 | 4519078 | ||
| BU147462.1 | - | 346 | ||||

ZMYND19
Genomic Map