siRNA Details
Gene Symbol: GINM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138785.3 | NP_620140.1 | 5 | 556 | ||
GenBank mRNA | AK289402.1 | - | 5 | 543 | ||
AK295690.1 | - | 4 | 485 | |||
AK308844.1 | - | 4 | 391 | |||
AK308994.1 | - | 2 | 212 | |||
AK311061.1 | - | 3 | 383 | |||
AK315729.1 | - | 5 | 553 | |||
AY358952.1 | - | 5 | 674 | |||
BC014320.1 | - | 5 | 560 | 3686673 |
