siRNA Details
Gene Symbol: GINM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138785.3 | NP_620140.1 | 8 | 957 | ||
GenBank mRNA | AK289402.1 | - | 8 | 944 | ||
AK315729.1 | - | 8 | 954 | |||
AY358952.1 | - | 8 | 1075 | |||
BC014320.1 | - | 8 | 961 | 3686673 |
