siRNA Details
| Gene Symbol: LINC01257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026670.2 | 2, 3 | 372 | |||
| GenBank mRNA | AL137487.1 | - | 2, 3 | 372 | ||
| BC112214.1 | - | 2, 3 | 347 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01257| Gene Symbol: LINC01257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026670.2 | 2, 3 | 372 | |||
| GenBank mRNA | AL137487.1 | - | 2, 3 | 372 | ||
| BC112214.1 | - | 2, 3 | 347 | |||