siRNA Details
| Gene Symbol: GRIN3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138690.2 | NP_619635.1 | 2 | 625 | ||
| GenBank mRNA | AY507106.1 | - | 2 | 625 | ||
| AY507107.1 | - | 2 | 625 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRIN3B| Gene Symbol: GRIN3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138690.2 | NP_619635.1 | 2 | 625 | ||
| GenBank mRNA | AY507106.1 | - | 2 | 625 | ||
| AY507107.1 | - | 2 | 625 | |||