siRNA Details
| Gene Symbol: CLNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052964.2 | NP_443196.2 | 12 | 756 | ||
| XM_011513775.2 | XP_011512077.1 | 12 | 987 | |||
| XM_017007684.1 | XP_016863173.1 | 12 | 987 | |||
| XM_017007685.1 | XP_016863174.1 | 12 | 987 | |||
| GenBank mRNA | AB032369.1 | - | 10 | 565 | ||
| AB110420.1 | - | 12 | 859 | |||
| AK093001.1 | - | 12 | 758 | |||
| BC029887.1 | - | 12 | 786 | |||
| BC137553.1 | - | 12 | 685 | 9021553 | ||
| BC137556.1 | - | 12 | 685 | 9021556 | ||

CLNK
Genomic Map