siRNA Details
| Gene Symbol: MRGPRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198923.2 | NP_944605.2 | 1 | 286 | ||
| GenBank mRNA | AB154410.1 | - | 1 | 286 | ||
| AY427820.1 | - | 1 | 286 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRGPRD| Gene Symbol: MRGPRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198923.2 | NP_944605.2 | 1 | 286 | ||
| GenBank mRNA | AB154410.1 | - | 1 | 286 | ||
| AY427820.1 | - | 1 | 286 | |||
Set Membership: |
|