siRNA Details
| Gene Symbol: MRPL53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053050.4 | NP_444278.1 | 3 | 535 | ||
| GenBank mRNA | BC012163.1 | - | 3 | 386 | 4622995 | |
| BG196300.1 | - | 3 | 519 | |||
| BQ688110.1 | - | 3 | 392 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRPL53| Gene Symbol: MRPL53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053050.4 | NP_444278.1 | 3 | 535 | ||
| GenBank mRNA | BC012163.1 | - | 3 | 386 | 4622995 | |
| BG196300.1 | - | 3 | 519 | |||
| BQ688110.1 | - | 3 | 392 | |||