siRNA Details
| Gene Symbol: SLC18B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052831.2 | NP_439896.1 | 3 | 363 | ||
| XM_017010236.1 | XP_016865725.1 | 3 | 554 | |||
| GenBank mRNA | AK124442.1 | - | 3 | 242 | ||
| AK289918.1 | - | 3 | 382 | |||
| AK293853.1 | - | 3 | 317 | |||
| BC069570.1 | - | 3 | 283 | 7262298 | ||

SLC18B1
Genomic Map