siRNA Details
| Gene Symbol: SLC18B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052831.2 | NP_439896.1 | 10 | 1166 | ||
| XM_017010236.1 | XP_016865725.1 | 10 | 1357 | |||
| GenBank mRNA | AJ420537.1 | - | 4 | 292 | ||
| AK124442.1 | - | 10 | 1045 | |||
| AK289918.1 | - | 10 | 1185 | |||
| AK293853.1 | - | 9 | 972 | |||
| BC069567.1 | - | 6 | 569 | |||
| BC069570.1 | - | 10 | 1086 | 7262298 | ||

SLC18B1
Genomic Map