siRNA Details
| Gene Symbol: LRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052972.2 | NP_443204.1 | 2 | 732 | ||
| GenBank mRNA | AB073607.1 | - | 2 | 744 | ||
| AF403428.1 | - | 2 | 732 | |||
| AK027382.1 | - | 2 | 744 | |||
| AK303861.1 | - | 3 | 693 | |||
| AK314506.1 | - | 2 | 744 | |||
| BC034389.1 | - | 2 | 733 | 5187278 | ||
| BC070198.1 | - | 2 | 789 | 4692618 | ||

LRG1
Genomic Map