siRNA Details
| Gene Symbol: DCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300854.1 | NP_001287783.1 | 4 | 449 | ||
| NM_053283.3 | NP_444513.1 | 4 | 449 | |||
| GenBank mRNA | AF144011.1 | - | 4 | 319 | ||
| AY590150.1 | - | 4 | 313 | |||
| BC062682.1 | - | 4 | 321 | 30369929 | ||
| BC069108.1 | - | 4 | 319 | 7216925 | ||
| BP313072.1 | - | 4 | 321 | |||
| EF503687.1 | - | 4 | 449 | |||
| EF503688.1 | - | 5 | 616 | |||
| EF503689.1 | - | 4 | 449 | |||

DCD
Genomic Map