siRNA Details
| Gene Symbol: ZNF354B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058230.2 | NP_478137.1 | 5 | 673 | ||
| GenBank mRNA | AK057737.1 | - | 5 | 626 | ||
| AK289667.1 | - | 5 | 583 | |||
| BC022190.2 | - | 5 | 671 | |||
| BC036466.1 | - | 5 | 671 | |||
| BC104777.1 | - | 4 | 504 | 8143780 | ||
| BC112111.1 | - | 4 | 504 | 8327579 | ||

ZNF354B
Genomic Map