siRNA Details
| Gene Symbol: BORCS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300742.1 | NP_001287671.1 | 2 | 83 | ||
| NM_058169.4 | NP_477517.1 | 2 | 474 | |||
| XM_011520551.2 | XP_011518853.1 | 2 | 117 | |||
| XM_011520553.2 | XP_011518855.1 | 2 | 467 | |||
| XM_017018778.1 | XP_016874267.1 | 2 | 467 | |||
| GenBank mRNA | AK075028.1 | - | 2 | 432 | ||
| AY037865.1 | - | 2 | 135 | |||
| BC013668.1 | - | 2 | 358 | 4154039 | ||
| BP260903.1 | - | 2 | 84 | |||

BORCS5
Genomic Map