siRNA Details
| Gene Symbol: MUCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058173.2 | NP_477521.1 | 4 | 385 | ||
| GenBank mRNA | AF414087.1 | - | 4 | 363 | ||
| AY359062.1 | - | 4 | 356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MUCL1| Gene Symbol: MUCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058173.2 | NP_477521.1 | 4 | 385 | ||
| GenBank mRNA | AF414087.1 | - | 4 | 363 | ||
| AY359062.1 | - | 4 | 356 | |||