siRNA Details
| Gene Symbol: BTBD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318189.1 | NP_001305118.1 | 15 | 1669 | ||
| NM_144587.3 | NP_653188.2 | 15 | 1666 | |||
| XM_011539239.2 | XP_011537541.1 | 14 | 2100 | |||
| XM_011539240.2 | XP_011537542.1 | 14 | 2075 | |||
| XM_011539241.2 | XP_011537543.1 | 13 | 2018 | |||
| XM_011539242.2 | XP_011537544.1 | 12 | 1899 | |||
| XM_017015636.1 | XP_016871125.1 | 14 | 2749 | |||
| XM_017015637.1 | XP_016871126.1 | 11 | 2374 | |||
| GenBank mRNA | AK058088.1 | - | 15 | 1656 | ||
| BC109261.1 | - | 15 | 1640 | 40025404 | ||
| BC109262.1 | - | 15 | 1640 | 40025414 | ||

BTBD16
Genomic Map