siRNA Details
| Gene Symbol: ZNF488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153034.2 | NP_694579.1 | 2 | 630 | ||
| XM_006717617.3 | XP_006717680.1 | 3 | 775 | |||
| XM_011539244.2 | XP_011537546.1 | 6 | 1068 | |||
| XM_017015642.1 | XP_016871131.1 | 2 | 679 | |||
| XM_017015643.1 | XP_016871132.1 | 4 | 2046 | |||
| GenBank mRNA | AK056666.1 | - | 2 | 644 | ||
| BC026895.1 | - | 3 | 512 | 4932309 | ||
| BC051323.1 | - | 2 | 630 | 6252494 | ||

ZNF488
Genomic Map