siRNA Details
Gene Symbol: FRA10AC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145246.4 | NP_660289.2 | 4 | 400 | ||
XM_005269515.4 | XP_005269572.1 | 5 | 497 | |||
XM_005269516.4 | XP_005269573.1 | 4 | 592 | |||
XM_011539259.2 | XP_011537561.1 | 4 | 365 | |||
XM_011539260.2 | XP_011537562.1 | 5 | 455 | |||
XM_017015656.1 | XP_016871145.1 | 4 | 365 | |||
XM_017015657.1 | XP_016871146.1 | 4 | 400 | |||
XM_017015658.1 | XP_016871147.1 | 5 | 500 | |||
XM_017015659.1 | XP_016871148.1 | 4 | 592 | |||
XM_017015660.1 | XP_016871149.1 | 5 | 457 | |||
XM_017015661.1 | XP_016871150.1 | 5 | 514 | |||
XM_017015662.1 | XP_016871151.1 | 4 | 365 | |||
GenBank mRNA | AJ431721.1 | - | 4 | 396 | ||
AJ431722.1 | - | 4 | 396 | |||
AJ431723.1 | - | 4 | 396 | |||
AJ431724.1 | - | 4 | 396 | |||
AJ431725.1 | - | 4 | 396 | |||
AK090955.1 | - | 4 | 400 | |||
AK308861.1 | - | 4 | 318 | |||
AK309016.1 | - | 4 | 513 | |||
BC018007.1 | - | 4 | 269 | 4278022 | ||
BM272089.1 | - | 4 | 384 | |||
BX648699.1 | - | 3 | 434 | |||
CN298116.1 | - | 4 | 379 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081075, AK077572, AK172241, AK017666.
Panther Classification:
Gene Ontology Categories:
