siRNA Details
| Gene Symbol: PDZD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173791.4 | NP_776152.1 | 2 | 1175 | ||
| XM_011539265.1 | XP_011537567.1 | 2 | 1148 | |||
| XM_011539266.2 | XP_011537568.1 | 2 | 1148 | |||
| GenBank mRNA | AK091746.1 | - | 2 | 1148 | ||
| AY211929.1 | - | 2 | 244 | |||
| BC028375.1 | - | 2 | 1137 | 4837939 | ||

PDZD8
Genomic Map