siRNA Details
| Gene Symbol: PDZD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173791.4 | NP_776152.1 | 3, 4 | 1331 | ||
| XM_005269518.3 | XP_005269575.1 | 2, 3 | 1181 | |||
| GenBank mRNA | AK091746.1 | - | 3, 4 | 1304 | ||
| AY211929.1 | - | 3, 4 | 400 | |||
| BC028375.1 | - | 3, 4 | 1293 | 4837939 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001033222, AK145020, BC141371, AK195576, AK041847, and others.

PDZD8
Genomic Map