siRNA Details
| Gene Symbol: CLRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152311.3 | NP_689524.1 | 2 | 496 | ||
| GenBank mRNA | BC029478.1 | - | 2 | 496 | 4733535 | |
| BM830163.1 | - | 2 | 249 | |||
| BX640823.1 | - | 1 | 776 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLRN3| Gene Symbol: CLRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152311.3 | NP_689524.1 | 2 | 496 | ||
| GenBank mRNA | BC029478.1 | - | 2 | 496 | 4733535 | |
| BM830163.1 | - | 2 | 249 | |||
| BX640823.1 | - | 1 | 776 | |||