siRNA Details
Gene Symbol: OR51F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004753.1 | NP_001004753.1 | 1 | 490 | ||
GenBank mRNA | BC136877.1 | - | 1 | 555 | 9020868 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR51F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004753.1 | NP_001004753.1 | 1 | 490 | ||
GenBank mRNA | BC136877.1 | - | 1 | 555 | 9020868 |