siRNA Details
| Gene Symbol: OR4X2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004727.1 | NP_001004727.1 | 1 | 814 | ||
| GenBank mRNA | BC136935.1 | - | 1 | 862 | 9020927 | |
| BC136936.1 | - | 1 | 862 | 9020928 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR4X2| Gene Symbol: OR4X2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004727.1 | NP_001004727.1 | 1 | 814 | ||
| GenBank mRNA | BC136935.1 | - | 1 | 862 | 9020927 | |
| BC136936.1 | - | 1 | 862 | 9020928 | ||