siRNA Details
| Gene Symbol: OR52K2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005172.2 | NP_001005172.2 | 1 | 917 | ||
| GenBank mRNA | BC137247.1 | - | 1 | 917 | 9021244 | |
| BC137248.1 | - | 1 | 917 | 9021245 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR52K2| Gene Symbol: OR52K2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005172.2 | NP_001005172.2 | 1 | 917 | ||
| GenBank mRNA | BC137247.1 | - | 1 | 917 | 9021244 | |
| BC137248.1 | - | 1 | 917 | 9021245 | ||