siRNA Details
| Gene Symbol: FAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008781.2 | NP_001008781.2 | 6 | 4263 | ||
| XM_017017178.1 | XP_016872667.1 | 8 | 4448 | |||
| XM_017017179.1 | XP_016872668.1 | 9 | 4670 | |||
| XM_017017180.1 | XP_016872669.1 | 8 | 4447 | |||
| XM_017017181.1 | XP_016872670.1 | 7 | 4378 | |||
| XM_017017182.1 | XP_016872671.1 | 8 | 4447 | |||
| XM_017017183.1 | XP_016872672.1 | 7 | 4378 | |||
| XM_017017184.1 | XP_016872673.1 | 8 | 4446 | |||
| XM_017017185.1 | XP_016872674.1 | 7 | 2274 | |||
| XM_017017186.1 | XP_016872675.1 | 7 | 1562 | |||
| XM_017017187.1 | XP_016872676.1 | 7 | 5920 | |||
| GenBank mRNA | CN295176.1 | - | 3 | 391 | ||

FAT3
Genomic Map