siRNA Details
| Gene Symbol: TMEM45B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138788.3 | NP_620143.1 | 4 | 578 | ||
| XM_005271400.4 | XP_005271457.1 | 5 | 703 | |||
| XM_011542586.2 | XP_011540888.1 | 5 | 747 | |||
| XM_011542588.2 | XP_011540890.1 | 5 | 698 | |||
| XM_017017188.1 | XP_016872677.1 | 6 | 853 | |||
| XM_017017189.1 | XP_016872678.1 | 6 | 869 | |||
| GenBank mRNA | AK098106.1 | - | 4 | 579 | ||
| AK290283.1 | - | 4 | 574 | |||
| AK292058.1 | - | 4 | 574 | |||
| BC016153.1 | - | 4 | 957 | 3841102 | ||

TMEM45B
Genomic Map