siRNA Details
| Gene Symbol: DNAJC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181706.4 | NP_859057.4 | 3, 4 | 368 | ||
| XM_017017216.1 | XP_016872705.1 | 3, 4 | 353 | |||
| GenBank mRNA | AK289665.1 | - | 3, 4 | 356 | ||
| AL833128.1 | - | 3, 4 | 358 | |||
| BC063804.1 | - | 3, 4 | 311 | 6474523 | ||
| CD685768.1 | - | 3, 4 | 386 | |||

DNAJC24
Genomic Map