siRNA Details
| Gene Symbol: DNAJC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181706.4 | NP_859057.4 | 4 | 399 | ||
| XM_017017216.1 | XP_016872705.1 | 4 | 384 | |||
| GenBank mRNA | AK289665.1 | - | 4 | 387 | ||
| AL833128.1 | - | 4 | 389 | |||
| BC036571.1 | - | 5 | 535 | 4793909 | ||
| BC044931.2 | - | 4 | 351 | |||
| BC063804.1 | - | 4 | 342 | 6474523 | ||
| CD685768.1 | - | 4 | 417 | |||

DNAJC24
Genomic Map