siRNA Details
| Gene Symbol: DNAJC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181706.4 | NP_859057.4 | 5 | 520 | ||
| XM_017017216.1 | XP_016872705.1 | 5 | 1293 | |||
| GenBank mRNA | AK289665.1 | - | 5 | 508 | ||
| AL833128.1 | - | 5 | 510 | |||
| BC044931.2 | - | 5 | 472 | |||
| BC063804.1 | - | 5 | 463 | 6474523 | ||
| CD685768.1 | - | 5 | 538 | |||

DNAJC24
Genomic Map