siRNA Details
| Gene Symbol: FAM186A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145475.1 | NP_001138947.1 | 1 | 290 | ||
| XM_006719231.3 | XP_006719294.1 | 1 | 301 | |||
| XM_011537890.2 | XP_011536192.1 | 1 | 301 | |||
| XM_011537892.1 | XP_011536194.1 | 1 | 30 | |||
| GenBank mRNA | AL833333.1 | - | 1 | 290 | ||
| BC054024.1 | - | 1 | 281 | |||

FAM186A
Genomic Map