siRNA Details
| Gene Symbol: TMEM132D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133448.2 | NP_597705.2 | 9 | 2499 | ||
| GenBank mRNA | AB061814.1 | - | 9 | 2500 | ||
| BC114626.1 | - | 9 | 2499 | 40082027 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM132D| Gene Symbol: TMEM132D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133448.2 | NP_597705.2 | 9 | 2499 | ||
| GenBank mRNA | AB061814.1 | - | 9 | 2500 | ||
| BC114626.1 | - | 9 | 2499 | 40082027 | ||